Skip to Content
Merck
All Photos(2)

Key Documents

SML3220

Sigma-Aldrich

GSK923295

≥95% (HPLC)

Synonym(s):

3-Chloro-N-{(1S)-2-[(N,N-dimethylglycyl)amino]-1-[(4-{8-[(1S)-1-hydroxyethyl]imidazo[1,2-a]pyridin-2-yl}phenyl)methyl]ethyl}-4-[(1-methylethyl)oxy]benzamide, GSK 923295, GSK 923295A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H38ClN5O4
CAS Number:
Molecular Weight:
592.13
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

GSK923295 is an allosteric inhibitor of CENP-E kinesin motor ATPase activity. It inhibits release of inorganic phosphate and stabilizes CENP-E motor domain interaction with microtubules, which causes failure of metaphase chromosome alignment and induces mitotic arrest. GSK923295 induces tumor cell apoptosis and tumor regression

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of the First Potent and Selective Inhibitor of Centromere-Associated Protein E: GSK923295
ACS Medicinal Chemistry Letters, 1(1), 30-34 (2010)
Zhen-Yu She et al.
Cell death discovery, 6, 25-25 (2020-05-01)
Kinesin-7 CENP-E is an essential kinetochore motor required for chromosome alignment and congression. However, the specific functions of CENP-E in the spermatogenic cells during spermatogenesis remain unknown. In this study, we find that CENP-E proteins are expressed in the spermatogonia
Rudolf Pisa et al.
Cell chemical biology, 27(7), 850-857 (2020-05-23)
Aberrant chromosome numbers in cancer cells may impose distinct constraints on the emergence of drug resistance-a major factor limiting the long-term efficacy of molecularly targeted therapeutics. However, for most anticancer drugs we lack analyses of drug-resistance mechanisms in cells with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service