Skip to Content
Merck
All Photos(1)

Key Documents

SML3181

Sigma-Aldrich

7,8-Dimethoxyperphenazine

≥98% (HPLC)

Synonym(s):

2-(4-(3-((3-chlorophenyl)(3,4-dimethoxyphenyl)amino)propyl)piperazin-1-yl)ethan-1-ol, 4-[3-[8-Chloro-2,3-dimethoxy-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£60.40
25 MG
£243.00

£60.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£60.40
25 MG
£243.00

About This Item

Empirical Formula (Hill Notation):
C23H30ClN3O3S
Molecular Weight:
464.02
UNSPSC Code:
12352200
NACRES:
NA.77

£60.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C2C(N(CCCN3CCN(CCO)CC3)C(C=C(C(OC)=C4)OC)=C4S2)=C1

InChI key

GKWVWOUUPIERSK-UHFFFAOYSA-N

Biochem/physiol Actions

7,8-Dimethoxyperphenazine is a cell penetrant inhibitor of UHM (U2AF homology motif) -ULM (U2AF ligand motif) interactions that interacts with the tryptophan binding pocket of different UHM domains including SPF45, PUF60 and U2AF. 7,8-Dimethoxyperphenazine inhibits early spliceosome assembly on pre-mRNA substrates in vitro.
cell penetrant inhibitor of UHM (U2AF homology motif) -ULM (U2AF ligand motif) interactions that interacts with the tryptophan binding pocket of UHM domains

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pravin Kumar Ankush Jagtap et al.
Nature communications, 11(1), 5621-5621 (2020-11-08)
Interactions between U2AF homology motifs (UHMs) and U2AF ligand motifs (ULMs) play a crucial role in early spliceosome assembly in eukaryotic gene regulation. UHM-ULM interactions mediate heterodimerization of the constitutive splicing factors U2AF65 and U2AF35 and between other splicing factors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service