Skip to Content
Merck
All Photos(1)

Key Documents

SML2864

Sigma-Aldrich

Loviride

≥98% (HPLC)

Synonym(s):

α-[(2-Acetyl-5-methylphenyl)amino]-2,6-dichloro-benzeneacetamide, (±)-2-(6-Acetyl-m-toluidino)-2-(2,6-dichlorophenyl)acetamide, R 89439

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16Cl2N2O2
CAS Number:
Molecular Weight:
351.23
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(c(ccc1)Cl)C(Nc2c(ccc(c2)C)C(=O)C)C(=O)N

InChI

1S/C17H16Cl2N2O2/c1-9-6-7-11(10(2)22)14(8-9)21-16(17(20)23)15-12(18)4-3-5-13(15)19/h3-8,16,21H,1-2H3,(H2,20,23)

InChI key

CJPLEFFCVDQQFZ-UHFFFAOYSA-N

Biochem/physiol Actions

Loviride is an orally available, potent and selective inhibitor of reverse transcriptase HIV-1 that inhibits HIV replication in cells. Also, loviride inhibits replication of HIV-2 and SIV in cells.
Orally available, potent and selective inhibitor of reverse transcriptase HIV-1 that inhibits HIV replication in cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Pauwels et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(5), 1711-1715 (1993-03-01)
In vitro evaluation of a large chemical library of pharmacologically acceptable prototype compounds in a high-capacity, cellular-based screening system has led to the discovery of another family of human immunodeficiency virus type 1 (HIV-1) inhibitors. Through optimization of a lead
Philippe Berben et al.
International journal of pharmaceutics, 537(1-2), 22-29 (2017-12-16)
In view of the increasing interest of pharmaceutical companies for cell- and tissue-free models to implement permeation into formulation testing, this study explored the capability of an artificial membrane insert system (AMI-system) as predictive tool to evaluate the performance of
M Witvrouw et al.
AIDS (London, England), 13(12), 1477-1483 (1999-08-28)
After the initial discovery of 1-(2-hydroxyethoxymethyl)-6-(phenylthio)thymine (HEPT) and tetrahydroimidazo[4,5,1-jk][1,4]benzodiazepin-2(1H)-one and thione (TIBO) derivatives, several other non-nucleoside reverse transcriptase (RT) inhibitors (NNRTI), including nevirapine (BI-RG-587), pyridinone derivatives (L-696,229 and L-697,661), delavirdine (U-90152), alpha-anilinophenylacetamides (alpha-APA) and various other classes of NNRTI have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service