Skip to Content
Merck
All Photos(1)

Key Documents

SML2314

Sigma-Aldrich

Celgosivir

≥98% (HPLC)

Synonym(s):

6-O-Butanoylcastanospermine, 6-O-Butyrylcastanospermine, BuCast, Butanoic acid (1S,6S,7S,8R,8aR)-octahydro-1,7,8-trihydroxy-6-indolizinyl ester, Butanoic acid [1S-(1α,6β,7α,8β,8aβ)]-octahydro-1,7,8-trihydroxy-6-indolizinyl ester, MDL 28,574, MDL 28574, MDL-28574

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£148.00
25 MG
£586.00

£148.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£148.00
25 MG
£586.00

About This Item

Empirical Formula (Hill Notation):
C12H21NO5
CAS Number:
Molecular Weight:
259.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

£148.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
under inert gas

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](OC(CCC)=O)CN2[C@@]([C@@H](O)CC2)([H])[C@H]1O

InChI

1S/C12H21NO5/c1-2-3-9(15)18-8-6-13-5-4-7(14)10(13)12(17)11(8)16/h7-8,10-12,14,16-17H,2-6H2,1H3/t7-,8-,10+,11+,12+/m0/s1

InChI key

HTJGLYIJVSDQAE-VWNXEWBOSA-N

Biochem/physiol Actions

Celgosivir is a prodrug of natural bicyclic iminosugar castanospermine that exhibits antiviral activity in vivo including HCV and dengue. Celgosivir is an inhibitor of α-glucosidases, which inhibits glycoprotein processing in cell culture. Apparently inhibition of α-glucosidases within infected cell prevents secretion of virus.
prodrug of natural bicyclic iminosugar castanospermine that exhibits antiviral activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C Sayce et al.
PLoS neglected tropical diseases, 10(3), e0004524-e0004524 (2016-03-15)
It has long been thought that iminosugar antiviral activity is a function of inhibition of endoplasmic reticulum-resident α-glucosidases, and on this basis, many iminosugars have been investigated as therapeutic agents for treatment of infection by a diverse spectrum of viruses
Ilaria Vicenti et al.
SLAS discovery : advancing life sciences R & D, 25(5), 506-514 (2020-03-19)
Practical cell-based assays can accelerate anti-Zika (ZIKV) and anti-dengue (DENV) virus drug discovery. We developed an immunodetection assay (IA), using a pan-flaviviral monoclonal antibody recognizing a conserved envelope domain. The final protocol includes a direct virus yield reduction assay (YRA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service