Skip to Content
Merck
All Photos(1)

Key Documents

SML1731

Sigma-Aldrich

A64 trifluoroacetate

≥98% (HPLC)

Synonym(s):

5-((6-Oxo-5-(6-(piperazin-1-yl)pyridin-3-yl)-1,6-dihydropyridin-3-yl)methylene)thiazolidine-2,4-dione trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£80.40

£80.40


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
£80.40

About This Item

Empirical Formula (Hill Notation):
C18H17N5O3S · xC2HF3O2
CAS Number:
Molecular Weight:
383.42 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

£80.40


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC=C(/C=C2SC(NC\2=O)=O)C=C1C3=CC=C(N4CCNCC4)N=C3.FC(F)(C(O)=O)F

Biochem/physiol Actions

A64 is a cell-permeable, ATP site-targeting pyridone thiazolidinedione compound that acts as a potent HIPK2-selective homeodomain-interacting protein kinase inhibitor (IC50 = 74 nM and 136 nM against HIPK2 and HIPK1, respectively; [ATP] = 100 μM), affecting MPF only at higher concentrations (by 35% at 5 μM; [ATP] = 10 μM) and exhibiting little potency toward CDK1 (IC50 >10 μM). A64 (1 μM) effectively prevents ER stresser tunicamycin (1 μg/mL) -induced HIPK2/JNK phosphorylation and cell death in HEK293 and primary rat motor neuron cultures. HIPK2 inhibition by A64 treatment is also efficacious in protecting rat motor neurons from death upon exogenous SOD1 G93A or TDP-43 expression. ff-target kinases revealed by kinome profiling include DYRK1A, CSNK2A2, DAPK2, DAPK1, PIM3, CSNK2A1, PIM1 (Kd = 8.8, 6.1, 6.1, 9.5, 3.7, 31, 37 nM, respectively) and DYRK1B (IC50 = 62 nM).
A64 is a cell-permeable, ATP site-targeting pyridone thiazolidinedione compound that acts as a potent HIPK2-selective homeodomain-interacting protein kinase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service