Skip to Content
Merck
All Photos(1)

Key Documents

SML0821

Sigma-Aldrich

UCD38B hydrochloride

≥98% (HPLC)

Synonym(s):

5’-Benzylglycinyl-amiloride, N-[6-Amino-5-[[(aminoiminomethyl)amino]carbonyl]-3-chloro-2-pyrazinyl]-glycine phenylmethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN7O3 · xHCl
CAS Number:
Molecular Weight:
377.79 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(NC(C(N=C1Cl)=C(N)N=C1CNC(OCC2=CC=CC=C2)=O)=O)=N.[H]Cl

InChI

1S/C15H16ClN7O3.ClH/c16-11-9(21-12(17)10(22-11)13(24)23-14(18)19)6-20-15(25)26-7-8-4-2-1-3-5-8;/h1-5H,6-7H2,(H2,17,21)(H,20,25)(H4,18,19,23,24);1H

InChI key

SFHYWDNJBRFZKW-UHFFFAOYSA-N

Biochem/physiol Actions

UCD38B is a cell permeable inhibitor of intracellular uPA (urokinase plasminogen activator) that is cytotoxic to proliferating and non-proliferating glioma cell lines.
UCD38B is a cell permeable inhibitor of intracellular uPA (urokinase plasminogen activator) that is cytotoxic to proliferating and non-proliferating glioma cell lines. UCD38B induces caspase-independent necrosis in U87 glioma MDA231 breast cancer cells. It appears that the necrotic pathway is mediated by AIF (apoptotic inducible factor).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service