Skip to Content
Merck
All Photos(1)

Key Documents

SMB01051

Sigma-Aldrich

Methyl jasminoside

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
£163.00

£163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
£163.00

About This Item

Empirical Formula (Hill Notation):
C27H32O13
CAS Number:
Molecular Weight:
564.54
UNSPSC Code:
12352205
NACRES:
NA.25

£163.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

564.54

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2OC=C([C@H](\C\2=C/COC(=O)\C=C/c3ccccc3)CC(=O)OC)C(=O)OC

InChI

1S/C27H32O13/c1-35-21(30)12-17-16(10-11-37-20(29)9-8-15-6-4-3-5-7-15)26(38-14-18(17)25(34)36-2)40-27-24(33)23(32)22(31)19(13-28)39-27/h3-10,14,17,19,22-24,26-28,31-33H,11-13H2,1-2H3/b9-8-,16-10+/t17-,19+,22+,23-,24+,26-,27-/m0/s1

InChI key

QBKMGAJNGCTGFA-OYDMIUNFSA-N

General description

Methyl jasminoside is a secoiridoid glucoside commonly found in plants from Oleaceace family (Jasminum amplexicaule and Jasminum odoratissimum).

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service