Skip to Content
Merck
All Photos(1)

Key Documents

SMB00355

Sigma-Aldrich

Herbacetin

≥98% (HPLC)

Synonym(s):

3,4′,5,7,8-Pentahydroxyflavone, 3,5,7,8-Tetrahydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7,8-Tetrahydroxyflavonol, 8-Hydroxykaempferol, Isoarticulatidin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
£166.00

£166.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
£166.00

About This Item

Empirical Formula (Hill Notation):
C15H10O7
CAS Number:
Molecular Weight:
302.24
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

£166.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=C2C(C(C(O)=C(C3=CC=C(O)C=C3)O2)=O)=C(O)C=C1O

InChI

1S/C15H10O7/c16-7-3-1-6(2-4-7)14-13(21)12(20)10-8(17)5-9(18)11(19)15(10)22-14/h1-5,16-19,21H

InChI key

ZDOTZEDNGNPOEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Herbacetin is a Met inhibitor and flavone found in the rhizomes of Rhodiola rosea.

Biochem/physiol Actions

Herbacetin has been reported to have potential in cancer therapeutics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service