Skip to Content
Merck
All Photos(1)

Key Documents

SIAL0488

Sigma-Aldrich

Sigma® cryogenic vial

capacity 2 mL, Internal thread, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121704
NACRES:
NB.22

material

colorless tube
polypropylene
polypropylene tube
round bottom (self-standing)

sterility

sterile; irradiated (Radiation Sterilization. Sterility Assurance Level: SAL 10^5)
sterile

feature

graduated
Internal thread
seal washer

packaging

pack of 10 × 50 ea
pack of 500 ea (10 x 50 ea)

manufacturer/tradename

Corning
Sigma-Aldrich

parameter

-196 °C min. temp.

capacity

2 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Cryogenic vials are manufactured from polypropylene to withstand temperatures to -196 °C.
  • Features a silicone washer for a secure seal.
  • Can be color-coded with inserts.
  • All self-standing vials have a special base design allowing them to be locked into the cryogenic rack and tray, for single-handed manipulation.

Legal Information

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service