Skip to Content
Merck
All Photos(1)

Key Documents

SCP0015

Sigma-Aldrich

α-MSH Peptide

≥90% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
£112.00

£112.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
£112.00

About This Item

Empirical Formula (Hill Notation):
C50H69N15O9
Molecular Weight:
1024.18
UNSPSC Code:
12352209
NACRES:
NA.32

£112.00


Please contact Customer Service for Availability

Product Name

[Asp5,D-Phe7,Lys10]-α MSH, Ac-Nle4 c amide 4-10,

Assay

≥90% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Nle-Asp-His-Phe-Arg-Trp-Lys-NH2

Application

α-melanocyte-stimulating hormone (MSH-α) is a neuroimmunomodulatory anti-inflammatory peptide with anti-microbial properties. D-Arg analogues such as [Asp5,D-Phe7,Lys10]-α MSH are used as α-MSH antagonists.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service