Skip to Content
Merck
All Photos(1)

Documents

SAE0023

Sigma-Aldrich

Alanine Racemase from Bacillus stearothermophilus

buffered aqueous solution, ≥10 IU/mg solid

Synonym(s):

Alanine Racemase from Bacillus stearothermophilus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Bacillus sp. (B. stearothermophilus)

Quality Level

form

buffered aqueous solution

specific activity

≥10 IU/mg solid

mol wt

Mr 78 kDa (2 subunits 39 kDa each)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Alanine racemase is used to convert L-alanine into D-alanine. Alanine racemase, from Sigma, has been used to isomerize L-[U-14C]alanine to a racemic mixture of L/D- [14C]alanine †.

Biochem/physiol Actions

Alanine racemase is involved in alanine, aspartate and D-alanine metabolism. 3-Fluoro-D-alanine and D-Cycloserine are known to inhibit alanine racemase.† Alanine racemase monomer is composed of two domains, an eight-stranded α/β barrel at the N-terminus and a C-terminal domain. The N-terminus includes residues 1-240, whereas the C-terminal comprises of the β-strand (residues 241-388). One molecule of pyridoxalphosphate (PLP) is present as the cofactor in each subunit.†

Unit Definition

One unit will convert 1.0 μmole of D-alanine to L-alanine per minute at pH 10.5 at 30 °C in a coupled assay system with L-alanine dehydrogenase.

Physical form

Aqueous solution containing 10 mM potassium phosphate

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service