Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702883

Sigma-Aldrich

Anti-SULT2A1 Antibody, clone 4A3H6, Rabbit Monoclonal

Synonym(s):

DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£331.00

£331.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
£331.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

£331.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

4A3H6, monoclonal

form

liquid

mol wt

34 kDa

species reactivity

rat, human

concentration

0.24 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

LLLSYEELKQDTGRTIEKICQFLGKTLEPEELNLILKNSSFQSMKENKMSNYSLLSVDYVVDKAQLLRKGVSGDWKNHFTVAQAEDFDKLFQEKMADLPRELFPWE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SULT2A1(6822)

General description

This gene encodes a member of the sulfotransferase family. Sulfotransferases aid in the metabolism of drugs and endogenous compounds by converting these substances into more hydrophilic water-soluble sulfate conjugates that can be easily excreted. This protein catalyzes the sulfation of steroids and bile acids in the liver and adrenal glands, and may have a role in the inherited adrenal androgen excess in women with polycystic ovary syndrome. [provided by RefSeq, Mar 2010]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 180-285 of human SULT2A1 (Q06520).

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service