Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702381

Sigma-Aldrich

Anti-BMP6 Antibody, clone 6N5S9, Rabbit Monoclonal

Synonym(s):

VGR, VGR1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
£331.00

£331.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
£331.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

£331.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

6N5S9, monoclonal

form

liquid

mol wt

42 kDa

species reactivity

rat, rat, human

concentration

0.50 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

DGPYDKQPFMVAFFKVSEVHVRTTRSASSRRRQQSRNRSTQSQDVARVSSASDYNSSELKTACRKHELYVSFQDLGWQDWIIAPKGYAANYCDGECSFPLN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP6(654)

General description

This gene encodes a secreted ligand of the TGF-beta (transforming growth factor-beta) superfamily of proteins. Ligands of this family bind various TGF-beta receptors leading to recruitment and activation of SMAD family transcription factors that regulate gene expression. The encoded preproprotein is proteolytically processed to generate each subunit of the disulfide-linked homodimer. This protein regulates a wide range of biological processes including iron homeostasis, fat and bone development, and ovulation. Differential expression of this gene may be associated with progression of breast and prostate cancer. Mutations in this gene may be associated with iron overload in human patients. [provided by RefSeq, Jul 2016]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 350-450 of human BMP6 (P22004).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service