Skip to Content
Merck
All Photos(3)

Key Documents

SAB5200658

Sigma-Aldrich

Monoclonal Anti-Dicer - Atto 594 antibody produced in mouse

clone S167-7, purified immunoglobulin

Synonym(s):

Anti-Double-strand-specific ribonuclease, Anti-HERNA, Anti-KIAA0928

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
£498.00

£498.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
£498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£498.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

Atto 594 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S167-7, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 215 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Dicer1(192119)

Immunogen

Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service