Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200030

Sigma-Aldrich

Anti-HOXD9 (Internal Region) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HOX4C, Anti-Homeobox D9, Anti-Hox-4.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells over expressing human HOXD9
indirect immunofluorescence: 2-5 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing human HOXD9
western blot: 1-2 μg/mL using lysates of HEK-293T cells over expressing human HOXD9

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXD9(3235)
mouse ... Hoxd9(15438)
rat ... Hoxd9(688999)

General description

There are 39 homeobox (HOX) genes in vertebrates. They are divided into four clusters (HOXA−HOXD), located on different chromosomes (7p15, 17q21.2, 12q13 and 2q31). Each cluster contains 9−11 member genes encoding relatively small gene products containing a highly conserved 60-amino-acid region (the homeobox), with DNA-binding activity that contributes to their activity as transcription factors. The HOX genes are evolutionarily conserved.

Application

Anti-HOXD9 (Internal Region) antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Homeobox (HOX) transcription factors control important development pathways involved embryo morphogenesis. HOX genes mainly participate in the formation of the body plan during embryonic development. Altered homeobox gene function or expression is implicated in the development of cancers such as leukemias or neoplasms of the breast, prostate, kidney, colon, skin, and brain. HOXD9 is involved in patterning of forelimb, axial skeleton and embryonic segmentation. It might promote cell migration, invasion and metastasis in hepatocellular carcinoma. HOXD9 was shown to be involved in the early stages of normal joint development and also in the pathologic process of arthritis.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone acetylation regulates the expression of HoxD9 transcription factor in endothelial progenitor cells
Iordache F, et al.
Romanian Journal of Morphology and Embryology, 56(1), 107-113 (2015)
Role of HOX genes in stem cell differentiation and Cancer
Bhatlekar S, et al.
Stem Cells International, 2018(1), 107-113 (2018)
HOXD9 promotes epithelial-mesenchymal transition and cancer metastasis by ZEB1 regulation in hepatocellular carcinoma
Lv X, et al.
Journal of Experimental & Clinical Cancer Research, 34(1), 133-133 (2015)
Expression of murine HOXD9 during embryonic joint patterning and in human T lymphotropic virus type I tax transgenic mice with arthropathy resembling rheumatoid arthritis.
Khoa ND, et al.
Arthritis and Rheumatism, 42(4), 686-696 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service