Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102838

Sigma-Aldrich

Anti-ZNF263 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FPM315, Anti-ZKSCAN12, Anti-Zinc finger protein 263

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

77 kDa

species reactivity

horse, bovine, dog, human, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF263(10127)

Immunogen

Synthetic peptide directed towards the middle region of human ZNF263

Biochem/physiol Actions

ZNF263 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers, 1 KRAB domain and 1 SCAN box domain. ZNF263 might play an important role in basic cellular processes as a transcriptional repressor.

Sequence

Synthetic peptide located within the following region: CHECGDSFSHSSNRIRHLRTHTGERPYKCSECGESFSRSSRLMSHQRTHT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service