Skip to Content
Merck
All Photos(4)

Documents

SAB2102397

Sigma-Aldrich

Anti-TCF7L2 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TCF4, Anti-Transcription factor 7-like 2 (T-cell specific, HMG-box)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCF7L2(6934)

Immunogen

Synthetic peptide directed towards the middle region of human TCF7L2

Biochem/physiol Actions

The TCL7L2 is a high mobility group (HMG) box-containing transcription factor implicated in blood glucose homeostasis. The study of Yi et al. suggested that TCL7L2 acts through regulation of proglucagon through repression of the proglucagon gene in enteroendocrine cells via the Wnt signaling pathway. The TCL7L2 gene product is a high mobility group (HMG) box-containing transcription factor implicated in blood glucose homeostasis. The study of Yi et al. (2005) [PubMed 15525634] suggested that TCL7L2 acts through regulation of proglucagon (MIM 138030) through repression of the proglucagon gene in enteroendocrine cells via the Wnt signaling pathway.[supplied by OMIM]. Sequence Note: This gene (TCF7L2, alias TCF-4) is distinct from TCF4. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-202 DA473655.1 1-202 203-251 Y11306.2 3-51 252-1252 AK225809.1 256-1256 1253-1305 Y11306.2 1053-1105 1306-1756 AK225809.1 1310-1760 1757-1811 Y11306.2 1557-1611 1812-2652 AK225809.1 1765-2605

Sequence

Synthetic peptide located within the following region: GGFRHPYPTALTVNASMSRFPPHMVPPHHTLHTTGIPHPAIVTPTVKQES

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service