Skip to Content
Merck
All Photos(2)

Documents

SAB1410748

Sigma-Aldrich

Anti-LAMA5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KIAA1907

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 74 kDa

species reactivity

human

technique(s)

proximity ligation assay: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA5(3911)

Related Categories

General description

Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined. (provided by RefSeq)

Immunogen

LAMA5 (AAH03355.1, 1 a.a. ~ 670 a.a) full-length human protein.

Sequence
MGVSLRDKKVHWVYQLGEAGPAVLSIDEDIGEQFAAVSLDRTLQFGHMSVTVERQMIQETKGDTVAPGAEGLLNLRPDDFVFYVGGYPSTFTPPPLLRFPGYRGCIEMDTLNEEVVSLYNFERTFQLDTAVDRPCARSKSTGDPWLTDGSYLDGTGFARISFDSQISTTKRFEQELRLVSYSGVLFFLKQQIQVFLLGGSRKRVLVRVERATVYSVEQDNDLELADAYYLGGVPPDQLPPSLRRLFPTGGSVRGCVKGIKALGKYVDLKRLNTTGVSAGCTADLLVGRAMTFHGHGFLRLALSNVAPLTGNVYSGFGFHSAQDSALLYYRASPDGLCQVSLQQGRVSLQLLRTEVKTQAGFADGAPHYVAFYSNATGVWLYVDDQLQQMKPHRGPPPELQPQPEGPPRLLLGGLPESGTIYNFSGCISNVFVQRLLGPQRVFDLQQNLGSVNVSTGCAPALQAQTPGLGPRGLQATARKASRRSRQPARHPACMLPPHLRTTRDSYQFGGSLSSHLEFVGILARHRNWPSLSMHVLPRSSRGLLLFTARLRPGSPSLALFLSNGHFVAQMEGLGTRLRAQSRQRSRPGRWHKVSVRWEKNRILLVTDGARAWSQEGPHRQHQGAEHPQPHTLFVGGLPASSHSSKLPTSQELHCLMWAWNWRCGPWQSPD

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service