Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407069

Sigma-Aldrich

Anti-CPSF4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CPSF30, NAR, NEB1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
£330.00

£330.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
£330.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

£330.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~27.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPSF4(10898)

General description

Inhibition of the nuclear export of poly(A)-containing mRNAs caused by the influenza A virus NS1 protein requires its effector domain. The NS1 effector domain functionally interacts with the cellular 30 kDa subunit of cleavage and polyadenylation specific factor 4, an essential component of the 3′ end processing machinery of cellular pre-mRNAs. In influenza virus-infected cells, the NS1 protein is physically associated with cleavage and polyadenylation specific factor 4, 30kD subunit. Binding of the NS1 protein to the 30 kDa protein in vitro prevents CPSF binding to the RNA substrate and inhibits 3′ end cleavage and polyadenylation of host pre-mRNAs. Thus the NS1 protein selectively inhibits the nuclear export of cellular, and not viral, mRNAs. Multiple alternatively spliced transcript variants that encode different isoforms have been described for this gene. (provided by RefSeq)

Immunogen

CPSF4 (AAH50738.1, 1 a.a. ~ 243 a.a) full-length human protein.

Sequence
MQEIIASVDHIKFDLEIAVEQQLGAQPLPFPGMDKSGAAVCEFFLKAACGKGGMCPFRHISGEKTVVCKHWLRGLCKKGDQCEFLHEYDMTKMPECYFYSKFGECSNKECPFLHIDPESKIKDCPWYDRGFCKHGPLCRHRHTRRVICVNYLVGFCPEGPSCKFMHPRFELPMGTTEQPPLPQQTQPPAKRTPQVIGVMQSQNSSAGNRGPRPLEQVTCYKCGEKGHYANRCTKGHLAFLSGQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service