Skip to Content
Merck
All Photos(1)

Key Documents

S2381

Sigma-Aldrich

DL-Stearoylcarnitine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H50ClNO4
CAS Number:
Molecular Weight:
464.12
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

Cl.CCCCCCCCCCCCCCCCCC(=O)OC(CC(O)=O)C[N](C)(C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nekadate and Blumberg
Cancer Research, 47, 6547-6547 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service