RAB1223
Equine PDGF-BB ELISA
for serum, plasma and cell culture supernatants
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
species reactivity
equine
technique(s)
ELISA: suitable
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
This ELISA antibody pair detects Equine PDGF-BB
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service