Recommended Products
material
polypropylene
feature
Holds 96 x 0.2 mL tubes
manufacturer/tradename
Heathrow Scientific HS23461D
technique(s)
PCR: suitable
size
127x99x30 mm
color
green
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Sturdy rack for working with 0.2 mL tubes. Accommodates the standard 8 or 12 tube strips. Perfect for pre- or post-PCR sampling applications. The tube wells are easily identified with imprinted numbers and letters. The rack features a removable hinged lid.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service