Key Documents
R1400
Texan reagent reservoir for multichannel pipettes
with lid, sterile, case of 100 ea (10 packs of 10ea)
Synonym(s):
multichannel pipette reservoir, reagent reservoir
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
material
V-bottom bottom
colorless
polypropylene
sterility
sterile
packaging
case of 100 ea (10 packs of 10ea)
manufacturer/tradename
Excel Scientific RSV-L175-S05
capacity
175 mL
General description
Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.
Legal Information
Texan is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What are the applications of this product?
    1 answer
    1. This product is a reservoir typically used as a holding tray or dish for use when transferring solutions or liquid reagents to multiwell plates or tubes at the bench. The v-shaped bottom allows for maximum pickup without needing both hands (one to tilt the tray). The product is most frequently used with multi-channel pipettors in liquid transfer as the image shows.
      Helpful?