Skip to Content
Merck
All Photos(1)

Key Documents

PZ0284

Sigma-Aldrich

PF-04753299

≥98%

Synonym(s):

α(R)-N-Hydroxy-α-methyl-α-(methylsulfonyl)-[1,1′-biphenyl]-4-butanamide, (+)-(2R)-4-(Biphenyl-4-yl)-N-hydroxy-2-methyl-2-(methylsulfonyl)butanamide, (2R)-4-Biphenyl-4-yl-N-hydroxy-2-methyl-2-(methylsulfonyl)butanamide, PF 04753299, PF-4753299

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£105.00
25 MG
£383.00

£105.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£105.00
25 MG
£383.00

About This Item

Empirical Formula (Hill Notation):
C18H21NO4S
CAS Number:
Molecular Weight:
347.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

£105.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(NO)[C@@](S(=O)(C)=O)(C)CCC(C=C1)=CC=C1C2=CC=CC=C2

InChI

1S/C18H21NO4S/c1-18(17(20)19-21,24(2,22)23)13-12-14-8-10-16(11-9-14)15-6-4-3-5-7-15/h3-11,21H,12-13H2,1-2H3,(H,19,20)/t18-/m1/s1

InChI key

GDYIQUFNICPYHF-GOSISDBHSA-N

Biochem/physiol Actions

PF-04753299 is a potent and selective inhibitor of LpxC (UDP-3-O-(R-3-hydroxymyristol)-N-acetylglucosamine deacetylase) that is effective in a murine of gram-negative bacteria infection model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service