Skip to Content
Merck
All Photos(1)

Key Documents

PZ0270

Sigma-Aldrich

PF-05190457

≥95% (HPLC)

Synonym(s):

1-[2-[(1R)-2,3-Dihydro-5-(6-methyl-4-pyrimidinyl)-1H-inden-1-yl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)-ethanone, 2-(2-Methylimidazo[2,1-b][1,3]thiazol-6-yl)-1-{2-[(1R)-5-(6-methylpyrimidin-4-yl)-2,3-dihydro-1H-inden-1-yl]-2,7-diazaspiro[3.5]non-7-ylethanone, PF 5190457, PF-5190457

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£102.00
25 MG
£378.00

£102.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£102.00
25 MG
£378.00

About This Item

Empirical Formula (Hill Notation):
C29H32N6OS
CAS Number:
Molecular Weight:
512.67
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

£102.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D +40 to +60°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CN2C(S1)=NC(CC(N3CCC4(CN([C@H](CCC5=C6)C5=CC=C6C7=NC=NC(C)=C7)C4)CC3)=O)=C2

InChI

1S/C29H32N6OS/c1-19-11-25(31-18-30-19)22-3-5-24-21(12-22)4-6-26(24)35-16-29(17-35)7-9-33(10-8-29)27(36)13-23-15-34-14-20(2)37-28(34)32-23/h3,5,11-12,14-15,18,26H,4,6-10,13,16-17H2,1-2H3/t26-/m1/s1

InChI key

ZIUDADZJCKGWKR-AREMUKBSSA-N

Biochem/physiol Actions

PF-05190457 is an orally available, potent and selective ghrelin receptor inverse agonist. PF-05190457 potently increases insulin secretion in glucose-stimulated human islets.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service