Skip to Content
Merck
All Photos(1)

Key Documents

PZ0263

Sigma-Aldrich

PF-3774076

≥98% (HPLC)

Synonym(s):

2-[(1R)-5-Chloro-2,3-dihydro-4-(methoxymethyl)-1H-inden-1-yl]-1H-imidazole, PF 3774076, PF-03774076

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£61.80
25 MG
£249.00

£61.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£61.80
25 MG
£249.00

About This Item

Empirical Formula (Hill Notation):
C14H15ClN2O
CAS Number:
Molecular Weight:
262.73
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

£61.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CC=C2C(CC[C@H]2C3=NC=CN3)=C1COC

InChI

1S/C14H15ClN2O/c1-18-8-12-10-2-3-11(14-16-6-7-17-14)9(10)4-5-13(12)15/h4-7,11H,2-3,8H2,1H3,(H,16,17)/t11-/m1/s1

InChI key

CMNAOKMSNCWOHS-LLVKDONJSA-N

Biochem/physiol Actions

PF-3774076 is a CNS penetrant, potent, selective, partial agonist at the human α1A adrenoceptor. PF-3774076 is selective over α1B and α1D adrenoceptors.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service