Skip to Content
Merck
All Photos(1)

Documents

PRI001

Sigma-Aldrich

PREADIPOCYTE ISOLATION KIT

Sufficient for 5 g of tissue

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352207
NACRES:
NA.75

usage

sufficient for 5 g tissue

packaging

pkg of 1 kit

storage condition

protect from light

technique(s)

protein purification: suitable
tissue culture: suitable

shipped in

wet ice

storage temp.

−20°C

General description

The Preadipocyte Isolation Kit provides the reagents and tools to isolate the stromal vascular fraction from adipose tissue. The resulting stromal vascular fraction is cultured on a tissue-culture plate, where preadipocytes adhere. The preadipocytes retain the ability to proliferate and differentiate into adipocytes when treated with differentiation-inducing components (DIF001, 3T3-L1 Differentiation Kit). The preadipocytes can be used to study the process of adipogenesis and the differentiated adipocytes can be used to study lipolysis, endocrine activity, cell-signaling, and metabolic dysfunction.

Application

Isolation of preadipocytes from adipose tissue.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service