P8248
Monoclonal Anti-Profilin antibody produced in mouse
~2 mg/mL, clone 2B8 (MAbPRF45a), purified immunoglobulin, buffered aqueous solution
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2B8 (MAbPRF45a), monoclonal
form
buffered aqueous solution
mol wt
antigen ~14 kDa
species reactivity
Arabidopsis thaliana
concentration
~2 mg/mL
technique(s)
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using total extracts of Arabidopsis flower
isotype
IgG1
shipped in
dry ice
storage temp.
−20°C
Gene Information
Arabidopsis thaliana ... PRF4(829055)
Looking for similar products? Visit Product Comparison Guide
General description
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service