N6260
4-Nitrophenyl phosphate di(2-amino-2-ethyl-1,3-propanediol) salt
≥97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
O2NC6H4OP(O)(OH)2 · (C5H14NO2)2
CAS Number:
Molecular Weight:
457.41
EC Number:
MDL number:
UNSPSC Code:
12352204
Recommended Products
Assay
≥97%
storage temp.
2-8°C
SMILES string
CCC(N)(CO)CO.CCC(N)(CO)CO.OP(O)(=O)Oc1ccc(cc1)[N+]([O-])=O
comparable product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service