Skip to Content
Merck
All Photos(1)

Key Documents

N6144

Sigma-Aldrich

[Ala5, β-Ala8]-α-Neurokinin Fragment 4-10

≥97% (HPLC)

Synonym(s):

Asp-Ala-Phe-Val-β-Ala-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H56N8O9S
CAS Number:
Molecular Weight:
764.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)CCNC(=O)C(NC(=O)C(Cc1ccccc1)NC(=O)C(C)NC(=O)C(N)CC(O)=O)C(C)C)C(N)=O

InChI

1S/C35H56N8O9S/c1-19(2)16-25(33(50)41-24(30(37)47)13-15-53-6)40-27(44)12-14-38-35(52)29(20(3)4)43-34(51)26(17-22-10-8-7-9-11-22)42-31(48)21(5)39-32(49)23(36)18-28(45)46/h7-11,19-21,23-26,29H,12-18,36H2,1-6H3,(H2,37,47)(H,38,52)(H,39,49)(H,40,44)(H,41,50)(H,42,48)(H,43,51)(H,45,46)

InChI key

AEABSNNHNNJKEX-UHFFFAOYSA-N

Gene Information

human ... TAC1(6863)

Amino Acid Sequence

Asp-Ala-Phe-Val-β-Ala-Leu-Met-NH2

Biochem/physiol Actions

NK-2 tachykinin receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Soejima et al.
Archives of oral biology, 44(6), 455-463 (1999-07-13)
Both substance P and neurokinin A are known as neurotransmitters of the submandibular ganglion cell. In this study, the effects of neurokinin (NK) receptor-subtype agonists on hamster submandibular ganglion cells were investigated using the whole-cell patch-clamp technique. Membrane currents evoked
S Evangelista et al.
Peptides, 11(2), 293-297 (1990-03-01)
We have developed a novel series of peptides, related to the NKA(4-10) sequence, in which substitution of selected amino acids determined variations in the affinity for the TK receptor subtypes. Subcutaneous pretreatment of rats with some peptides of this series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service