Skip to Content
Merck
All Photos(1)

Key Documents

N2002

Sigma-Aldrich

4-Nitrophenyl dodecanoate

Synonym(s):

4-Nitrophenyl laurate, Dodecanoic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27NO4
CAS Number:
Molecular Weight:
321.41
Beilstein:
1889084
EC Number:
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C18H27NO4/c1-2-3-4-5-6-7-8-9-10-11-18(20)23-17-14-12-16(13-15-17)19(21)22/h12-15H,2-11H2,1H3

InChI key

YNGNVZFHHJEZKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service