Skip to Content
Merck
All Photos(1)

Key Documents

N1001

Sigma-Aldrich

1-Naphthyl valerate

liquid

Synonym(s):

Pentanoic acid 1-naphthyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

color

colorless to pale yellow

storage temp.

−20°C

SMILES string

CCCCC(=O)Oc1cccc2ccccc12

InChI

1S/C15H16O2/c1-2-3-11-15(16)17-14-10-6-8-12-7-4-5-9-13(12)14/h4-10H,2-3,11H2,1H3

InChI key

GRGNAHUPACBFOF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Haenig et al.
Cell reports, 32(7), 108050-108050 (2020-08-20)
Interactome maps are valuable resources to elucidate protein function and disease mechanisms. Here, we report on an interactome map that focuses on neurodegenerative disease (ND), connects ∼5,000 human proteins via ∼30,000 candidate interactions and is generated by systematic yeast two-hybrid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service