MTCC2230AM
MTC™ Bio ScrewSeal™ Microcentrifuge Tubes
self standing, amber, capacity 2.0 mL, pkg of 1000 ea (10 x bags 100 ea)
Synonym(s):
conical tube, plastic tube
All Photos(1)
About This Item
Recommended Products
material
amber
conical
high-density polyethylene cap
polypropylene
screw top cap
sterility
sterile
feature
graduation
packaging
pkg of 1000 ea (10 x bags 100 ea)
manufacturer/tradename
MTC™ Bio C2230-AM
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
12.8 mm × 47.4 mm
Related Categories
General description
Screw-cap microtubes, 2.0mL, amber color, with O-ring, sterile, molded graduations, amber caps attached, 100 per bag, 1000/case
ScrewSeal™ screw-cap microtubes are made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leak-proof seal from -90°C to +121°C. Sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g.
Features and Benefits
- Ultra-clear polypropylene
- Molded or non-graduated options
- Leak-proof O-ring seal
- Light-blocking amber available
- Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
ScrewSeal is a trademark of MTC Bio