Skip to Content
Merck
All Photos(1)

Key Documents

M5648

Sigma-Aldrich

(S)-(+)-2-Methoxy-2-phenylethanol

liquid

Synonym(s):

β-Methoxyphenethyl alcohol, (+)-β-Methoxyphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CH2OH
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352000
Pricing and availability is not currently available.

form

liquid

color

light yellow

refractive index

n20/D 1.52 (lit.)

bp

65 °C/0.1 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

CO[C@H](CO)c1ccccc1

InChI

1S/C9H12O2/c1-11-9(7-10)8-5-3-2-4-6-8/h2-6,9-10H,7H2,1H3/t9-/m1/s1

InChI key

JDTUPLBMGDDPJS-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

May darken on storage

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service