Skip to Content
Merck
All Photos(1)

Key Documents

M3699

Sigma-Aldrich

Methedrone hydrochloride

≥98% (HPLC)

Synonym(s):

Methedrone hydrochloride, 1-(4-Methoxy-phenyl)-2-methylamino-propan-1-one hydrochloride, 4-Methoxymethcathinone hydrochloride, Methoxyephedrone hydrochloride, Methoxyphedrine hydrochloride, PMMC hydrochloride, bk-PMMA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2·HCl
CAS Number:
Molecular Weight:
229.70
UNSPSC Code:
41116107
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to off-white

solubility

H2O: >20 mg/mL

storage temp.

room temp

InChI

1S/C11H15NO2.ClH/c1-8(12-2)11(13)9-4-6-10(14-3)7-5-9;/h4-8,12H,1-3H3;1H

InChI key

QIJFAKGYWIXUDG-UHFFFAOYSA-N

Biochem/physiol Actions

Methedrone is a synthetic stimulant, an analog of Cathinone and Methamphetamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Gibbons et al.
Bioorganic & medicinal chemistry letters, 20(14), 4135-4139 (2010-06-15)
'Legal highs' are compounds, plant or fungal material which can be readily bought from the internet without legal restriction and the single chemicals may be structurally related to illegal drugs of abuse such as the amphetamines. Several recent deaths in
Priscilla B Pail et al.
Neurotoxicology, 50, 71-80 (2015-08-10)
Mephedrone and methedrone are cathinone-related compounds, which act as non-selective substrates for monoamine transporters, facilitating a neurotransmitter release. We compared the acute pharmacological effects of mephedrone and methedrone, attempting to further evaluate the action mechanisms of methedrone by responsibly and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service