Skip to Content
Merck
All Photos(1)

Documents

M2324

Sigma-Aldrich

ML-3H2

Synonym(s):

5-Chloro-7-(diethylaminomethyl)quinolin-8-ol, 5-Chloro-7-[(diethylamino)methyl]-8-quinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O
CAS Number:
Molecular Weight:
264.75
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.41

form

solid

solubility

DMSO: 20 mg/mL
soluble

storage temp.

2-8°C

SMILES string

CCN(CC)Cc1cc(Cl)c2cccnc2c1O

InChI

1S/C14H17ClN2O/c1-3-17(4-2)9-10-8-12(15)11-6-5-7-16-13(11)14(10)18/h5-8,18H,3-4,9H2,1-2H3

InChI key

QDFFXEIFWNNJJU-UHFFFAOYSA-N

Biochem/physiol Actions

ML-3H2 is an allosteric hexamer-stabilizing inhibitor of human porphobilinogen synthase (PBGS;ALAD)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service