Skip to Content
Merck
All Photos(1)

Key Documents

M0376

Sigma-Aldrich

L-Methionine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
199.70
Beilstein:
3913214
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

151-153 °C (lit.)

storage temp.

−20°C

SMILES string

Cl[H].COC(=O)[C@@H](N)CCSC

InChI

1S/C6H13NO2S.ClH/c1-9-6(8)5(7)3-4-10-2;/h5H,3-4,7H2,1-2H3;1H/t5-;/m0./s1

InChI key

MEVUPUNLVKELNV-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vertuani Silvia et al.
European journal of medicinal chemistry, 50, 383-392 (2012-03-06)
Following the recent output of scientific publications in the matter of synergic activity between different antioxidants, we have undertaken the present study with the aim to synthesize new molecules with radical-scavengers activity based on the conjugation of bioactive portions (i.e.
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service