Skip to Content
Merck
All Photos(1)

Key Documents

L5753

Sigma-Aldrich

Linoleoyl chloride

≥99%, liquid

Synonym(s):

9,12-Octadecadienoyl chloride, (9Z,12Z)- (9CI, ACI)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H31OCl
CAS Number:
Molecular Weight:
298.89
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

plant

Assay

≥99%

form

liquid

density

0.93 g/mL at 25 °C (lit.)

functional group

chloro

lipid type

omega FAs

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(Cl)=O

InChI

1S/C18H31ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3/b7-6-,10-9-

InChI key

FBWMYSQUTZRHAT-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Linoleoyl chloride is used as a reagent for the synthesis of fatty acyl glyceric acids such as dilinoleoyl-D-glyceric acid

Packaging

Sealed ampule.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hiroshi Habe et al.
Journal of oleo science, 60(6), 327-331 (2011-05-25)
Previously, Lešová et al. reported the isolation and identification of metabolite OR-1, showing antitrypsin activity, produced during fermentation by Penicillium funiculosum. The structure of OR-1 was a mixture of glyceric acid (GA), esterified with C(14)-C(18) fatty acids, and oleic acid
Jing Xie et al.
BioMed research international, 2013, 710502-710502 (2013-07-19)
A novel antisense oligonucleotide (ASO) carrier, polyethylenimine conjugated to linoleic acid (PEI-LA), was synthesized and evaluated for delivery of LOR-2501 to tumor cells. LOR-2501 is an ASO targeting ribonucleotide reductase R1 subunit (RRM1). In this study, PEI-LA was synthesized by
Ahmed Abu-Fayyad et al.
International journal of pharmaceutics, 528(1-2), 463-470 (2017-06-20)
Gemcitabine is the first line therapy for pancreatic cancer. It is, however, extensively metabolized to the inactive form by deamination enzymatic reaction. Conjugation of gemcitabine with fatty acids on its 4-amino group was found to protect it from deamination deactivation
Shun Sato et al.
Journal of oleo science, 60(9), 483-487 (2011-08-20)
A novel derivative of glyceric acid (GA), dilinoleoyl-D-glyceric acid (LA₂-DGA), was synthesized from D-GA calcium salt and linoleoyl chloride and evaluated for cytotoxicity. The D-GA calcium salt was first reacted with 4-methoxybezylchloride, and the resulting compound was esterified with linoleoyl
Siddabasave Gowda B Gowda et al.
Antioxidants (Basel, Switzerland), 9(5) (2020-05-14)
Branched fatty acid esters of hydroxy fatty acids (FAHFAs) are a recently discovered class of biologically active lipids with anti-inflammatory and anti-diabetic properties. Despite the possible link between endogenous FAHFA levels and nuclear factor erythroid 2-related factor 2 (Nrf2), their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service