Skip to Content
Merck
All Photos(1)

Key Documents

L4275

Sigma-Aldrich

Linseed oil

suitable for insect cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

technique(s)

cell culture | insect: suitable

refractive index

n20/D 1.4795 (lit.)

density

0.93 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ebrahimi et al.
BioMed research international, 2013, 194625-194625 (2013-03-14)
This study was conducted to determine the effects of feeding oil palm frond silage based diets with added linseed oil (LO) containing high α -linolenic acid (C18:3n-3), namely, high LO (HLO), low LO (LLO), and without LO as the control
Joana R N Lemos et al.
Nutrition research (New York, N.Y.), 32(12), 921-927 (2012-12-19)
Malnutrition and chronic inflammation in dialysis patients negatively impact their survival prognosis, and nutrients, such as omega-3 oils, are postulated to reduce proinflammatory response. In this randomized, double-blind, multicenter, placebo-controlled trial, we investigated the effects of flaxseed oil (FO) on
Chunhua Luo et al.
Acta biomaterialia, 9(2), 5226-5233 (2012-09-15)
Totally bio-based thermosetting polymers which are comparable to synthetic polyester thermosets have been prepared from copolymerization of condensed tannin-fatty acid esters with vegetable oils. Oxidative copolymerization of tannin linoleate/acetate mixed esters with linseed oil and tung oil produced polymer films
Ilaria Bonaduce et al.
PloS one, 7(11), e49333-e49333 (2012-11-21)
This paper presents an analytical investigation of paint reconstructions prepared with linseed oil that have undergone typical 19th century treatments in preparation for painting. The oil was mechanically extracted from the same seed lot, which was then processed by various
N Renu et al.
Acta gastro-enterologica Belgica, 75(3), 331-335 (2012-10-23)
The present study was undertaken to elucidate the effect of Linum usitatissimum fixed oil on experimental esophagitis in albino rats. Group of rats (n = 6), treated with vehicle control (0.9% NaCl, 3 mL/kg, i.p.) or L. usitatissimum fixed oil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service