Skip to Content
Merck
All Photos(1)

Key Documents

L2899

Sigma-Aldrich

Leukotriene E4 methyl ester

~100 μg/mL in methanol: water (3:1) (containing 17 mM ammonium acetate buffer, pH 5.4), ~97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

~97% (HPLC)

concentration

~100 μg/mL in methanol: water (3:1) (containing 17 mM ammonium acetate buffer, pH 5.4)

shipped in

dry ice

storage temp.

−20°C

Packaging

Packaged under Argon.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service