Journal of applied microbiology, 111(6), 1371-1380 (2011-09-08)
The aim of this study is to improve exoinulinase production by expression of a cloned exoinulinase gene inuA1 (GenBank accession no. JF961344) from Penicillium janthinellum strain B01 in Pichia pastoris. A full-length cDNA of exoinulinase gene (inuA1) was cloned from
Though some genetic features of lactobacillar fructan hydrolases were elucidated, information about their enzymology or mutational analyses were scarce. Lactobacillus casei IAM1045 exhibits extracellular activity degrading inulin. After partial purification of the inulin-degrading protein from the spent culture medium, several
Anais da Academia Brasileira de Ciencias, 84(2), 443-454 (2012-05-29)
Inulinase (β-2,1-D- fructan fructanohydrolase), EC 3.2.1.7, targets the β-2,1 linkage of inulin, a polyfructan consisting of linear β-2,1 linked fructose, and hydrolyzes it into fructose. This use provides an alternative to produce fructose syrup through the hydrolysis of inulin. The
Bioprocess and biosystems engineering, 35(3), 383-388 (2011-08-13)
An experimental design was carried out to evaluate the effect of the concentrations of sodium alginate, glutaraldehyde and activated coal on the immobilization of inulinase from Kluyveromyces marxianus NRRL Y-7571. The experimental condition of 20 g/L of sodium alginate, 50 mL/L of
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 27(7), 1032-1039 (2011-10-25)
Ethanol fermentation from Jerusalem artichoke tubers by recombinant Saccharomyces cerevisiae strains expressing the inulinase gene (inu) from Kluyveromyces marxianus was investigated. The inu native and pgk promoters were used to drive the expression of the inu gene, and the inulinase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.