I6272
Magnesium ionophore IV
Synonym(s):
ETH 7025, N,N′,N′′-Tris[3-(heptylmethylamino)-3-oxopropionyl]-8,8′-iminodioctylamine
About This Item
storage temp.
2-8°C
SMILES string
CCCCCCCN(C)C(=O)CC(=O)NCCCCCCCCN(CCCCCCCCNC(=O)CC(=O)N(C)CCCCCCC)C(=O)CC(=O)N(C)CCCCCCC
InChI
1S/C49H94N6O6/c1-7-10-13-22-29-36-52(4)46(58)41-44(56)50-34-27-20-16-18-25-32-39-55(49(61)43-48(60)54(6)38-31-24-15-12-9-3)40-33-26-19-17-21-28-35-51-45(57)42-47(59)53(5)37-30-23-14-11-8-2/h7-43H2,1-6H3,(H,50,56)(H,51,57)
InChI key
JYBGTBZZVHYIJI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Physical form
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service