Assay
~90% (TLC)
storage temp.
−20°C
SMILES string
[K+].[K+].[K+].[K+].[K+].Cc1ccc(N(CC([O-])=O)CC([O-])=O)c(OCCOc2cc(ccc2N(CC([O-])=O)CC([O-])=O)-c3cc4ccc(cc4[nH]3)C([O-])=O)c1
Biochem/physiol Actions
Cell impermeable fluorescent probe for Ca2+; lower affinity than FURA 2; gives a large shift in fluorescent emission from 482 nm to 398 nm upon Ca2+ binding.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Circulation research, 67(2), 415-425 (1990-08-01)
In studies that attempt to measure intracellular calcium [( Ca2+]i) in the intact heart with the calcium indicator indo 1-AM, a fundamental assumption is that the signals report changes in myocyte [Ca2+]i. We studied isolated perfused rabbit hearts loaded with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service