Skip to Content
Merck
All Photos(1)

Key Documents

I2523

Sigma-Aldrich

D-myo-Inositol 1-monophosphate bis(cyclohexylammonium) salt

from soybean L-α-phosphatidylinositol, ≥93%, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O9P · 2C6H13N
Molecular Weight:
458.48
Beilstein:
3923085
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

soybean (L-α-phosphatidylinositol)

Assay

≥93%

form

lyophilized powder

impurities

inositol 2-monophosphate, trace

storage temp.

−20°C

SMILES string

NC1CCCCC1.O[C@H]2[C@H](O)[C@@H](O)[C@H](OP(O)(O)=O)[C@H](O)[C@@H]2O

InChI

1S/C6H13N.C6H13O9P/c7-6-4-2-1-3-5-6;7-1-2(8)4(10)6(5(11)3(1)9)15-16(12,13)14/h6H,1-5,7H2;1-11H,(H2,12,13,14)/t;1-,2-,3+,4-,5-,6-/m.1/s1

InChI key

CIDVDWGHBSYGCB-QNNMCHRWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service