Key Documents
HS120247
Wide mouth wash bottle
capacity 500 mL, yellow PE-LD bottle
Select a Size
1 EACH
£30.80
£30.80
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
£30.80
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
£30.80
Please contact Customer Service for Availability
material
polypropylene cap
yellow PE-LD bottle
feature
label no label
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120247
capacity
500 mL
General description
Color-coded tops for easy identification
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
  • Wide bottle mouth makes filling easier
  • Color-coding for user, application or location identification
  • Bottle is made from LDPE, with a PP closure
  • 500 mL bottle size,
  • Supplied in packs of 6
Choose from one of the most recent versions:
Certificates of Analysis (COA)