Skip to Content
Merck
All Photos(1)

Key Documents

HPA014014

Sigma-Aldrich

Anti-GPR31 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Probable G-protein coupled receptor 31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

NAGIIRALQKRLREPEKQPKLQRAQA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR31(2853)

General description

The gene GPR31 (G protein-coupled receptor 31) encodes a member of the GPCR family. The proteins belonging to this family are characterized by the presence of seven transmembrane domains. The encoded protein is around 319 amino acid long and belongs to the rhodopsin-like group A subfamily. The gene is mapped to human chromosome 6q27.

Immunogen

Probable G-protein coupled receptor 31 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene GPR31 (G protein-coupled receptor 31) encodes an orphan G protein-coupled receptor that binds to 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE), a human 12-lipoxygenase-derived product. It is localized to the plasma membrane and is also referred to as 12-HETER (12-(S)-HETE receptor). It also binds to 12-(S)-HETE hydroxyl fatty acids with high affinity. The binding partner, 12-(S)-HETE, functions in several processes including cytoskeleton remodeling, stimulation of cells to secrete proteinases and vascular endothelial growth factor resulting in an angiogenic response. The binding of 12-(S)-HETE to GPR31 activates MEK (MAPK/ERK kinase), ERK1/2 (extracelular signal regulated kinase), and NFκB (nuclear factor) signaling molecules. This ligand-receptor complex is also involved in the regulation of tumor cell invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72744

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth V Honn et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(6), 2360-2369 (2016-03-12)
Previously we identified and deorphaned G-protein-coupled receptor 31 (GPR31) as the high-affinity 12(S)-hydroxyeicosatetraenoic acid [12(S)-HETE] receptor (12-HETER1). Here we have determined its distribution in prostate cancer tissue and its role in prostate tumorigenesis using in vitro and in vivo assays.
Yande Guo et al.
The Journal of biological chemistry, 286(39), 33832-33840 (2011-06-30)
Hydroxy fatty acids are critical lipid mediators involved in various pathophysiologic functions. We cloned and identified GPR31, a plasma membrane orphan G protein-coupled receptor that displays high affinity for the human 12-lipoxygenase-derived product 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE). Thus, GPR31 is named
A Zingoni et al.
Genomics, 42(3), 519-523 (1997-06-15)
The screening of a human genomic library with a chemokine receptor-like probe allowed us to obtain a putative member of the G protein-coupled receptor gene (GPCR) family, designated GPR31. Its deduced amino acid sequence encodes a polypeptide of 319 amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service