Skip to Content
Merck
All Photos(5)

Key Documents

HPA008477

Sigma-Aldrich

Anti-PRSS21 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ESP-1 antibody produced in rabbit, Anti-Eosinophil serine protease 1 antibody produced in rabbit, Anti-Serine protease 21 antibody produced in rabbit, Anti-Testisin precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

CFETYSDLSDPSGWMVQFGQLTSMPSFWSLQAYYTRYFVSNIYLSPRYLGNSPYDIALVKLSAPVTYTKHIQPICLQASTFEFENRTDCWVTGWGYIKEDEALPSPHTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRSS21(10942)

General description

Serine protease 21 (PRSS21) is localized to premeiotic testicular germ cells. The gene encoding this protein is present on the short arm of chromosome 16p13.3. It consists of an N-terminal signal sequence of 27 amino acids, a 251 amino-acid catalytic domain with trypsin-like specificity and a C-terminal hydrophobic region which acts as a membrane anchor.

Immunogen

Testisin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Serine protease 21 (PRSS21) functions as a tumor suppressor for testicular cancer. It has been shown that the trypsin-like protease activity of PRSS21 is essential for the process of sperm penetration through the cumulus matrix and zona pellucida of the oocyte in vitro. Hence this trypsin-like activity is important for fertilization in vitro but not necessarily in vivo. PRSS21 also contributes to the epididymal sperm maturation and the ability of the sperms to fertilize the oocyte.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70760

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Hooper et al.
Biochimica et biophysica acta, 1492(1), 63-71 (2000-09-27)
Testisin is a recently identified human serine protease expressed by premeiotic testicular germ cells and is a candidate tumor suppressor for testicular cancer. Here, we report the characterization of the gene encoding testisin, designated PRSS21, and its localization on the
A L Scarman et al.
European journal of biochemistry, 268(5), 1250-1258 (2001-03-07)
The recently characterized human serine protease, Testisin, is expressed on premeiotic testicular germ cells and is a candidate type II tumor suppressor for testicular cancer. Here we report the cloning, characterization and expression of the gene encoding mouse Testisin, Prss21.
Sarah Netzel-Arnett et al.
Biology of reproduction, 81(5), 921-932 (2009-07-03)
An estimated 25%-40% of infertile men have idiopathic infertility associated with deficient sperm numbers and quality. Here, we identify the membrane-anchored serine protease PRSS21, also known as testisin, to be a novel proteolytic factor that directs epididymal sperm cell maturation
Natsuko Kawano et al.
Biology of reproduction, 83(3), 359-369 (2010-05-21)
Although sperm serine protease and proteasome have long been believed to play an important role in the fertilization process, the molecular mechanism is still controversial. In this study, we have produced double-knockout mice lacking two sperm serine proteases, ACR and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service