Skip to Content
Merck
All Photos(5)

Key Documents

H2769

Sigma-Aldrich

Anti-Hyperpolarization-Activated Cation Channel HCN2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 ML
£260.00

£260.00


Please contact Customer Service for Availability


Select a Size

Change View
0.05 ML
£260.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

£260.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1:200-1:300 using rat brain tissue

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HCN2(610)
mouse ... Hcn2(15166)
rat ... Hcn2(114244)

Related Categories

Immunogen

synthetic peptide corresponding to amino acids 147-161 of human HCN2 (with additional N-terminal cysteine). The epitope is identical in mouse.

Physical form

Lyophilized at 0.3 mg/mL from Phosphate Buffered Saline, pH 7.4, containing 1% Bovine Serum Albumin, and 0.05% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service