Skip to Content
Merck
All Photos(1)

Key Documents

H2006

Sigma-Aldrich

1-Hydroxybenzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3O
CAS Number:
Molecular Weight:
135.12
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

156-159 °C (lit.)

SMILES string

On1nnc2ccccc12

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits racemization in peptide syntheses using DCC as condensing agent.

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Expl. 1.3 - Eye Irrit. 2

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 1

Flash Point(F)

293.0 °F - Pensky Martens closed cup

Flash Point(C)

145 °C - Pensky Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[A new method for synthesis of peptides: activation of the carboxyl group with dicyclohexylcarbodiimide using 1-hydroxybenzotriazoles as additives].
W König et al.
Chemische Berichte, 103(3), 788-798 (1970-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service