Skip to Content
Merck
All Photos(1)

Key Documents

H1265

Sigma-Aldrich

cis-3-Hexen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1719712
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3.45 (vs air)

refractive index

n20/D 1.44 (lit.)

bp

156-157 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

CC\C=C/CCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h3-4,7H,2,5-6H2,1H3/b4-3-

InChI key

UFLHIIWVXFIJGU-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service